ENA061488120 Enamine

3-{2-amino-5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-6-yl}propanoic acid

  • MDL number MFCD07323325
  • Cas Number : 847744-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13764
Availability details for
Quantity
  • Availability
  • Add to Cart