ENA061488144 Enamine

2-[3-mercapto-5-(3-methylphenyl)-4H-1,2,4-triazol-4-yl]acetamide

  • MDL number MFCD07324829
  • Cas Number : 847744-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13813
Availability details for
Quantity
  • Availability
  • Add to Cart