ENA061488149 Enamine

1-[(2,5-difluorophenyl)sulfonyl]piperazine

  • MDL number MFCD07323193
  • Cas Number : 847837-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13824
Availability details for
Quantity
  • Availability
  • Add to Cart