ENA061488153 Enamine

(2E)-3-(4-azepan-1-ylphenyl)acrylic acid

  • MDL number MFCD07323190
  • Cas Number : 847837-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13831

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13831
Availability details for
Quantity
  • Availability
  • Add to Cart