ENA061488156 Enamine

ethyl 2-(2,5-dichlorothiophen-3-yl)-2-oxoacetate

  • MDL number MFCD07323196
  • Cas Number : 32766-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13834
Availability details for
Quantity
  • Availability
  • Add to Cart