ENA061488169 Enamine

methyl 4-(2,4-dimethoxyphenyl)-2,4-dioxobutanoate

  • MDL number MFCD07323955
  • Cas Number : 39757-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13855
Availability details for
Quantity
  • Availability
  • Add to Cart