ENA061488175 Enamine

2-[3-(2-chloroacetyl)-2,5-dimethyl-1H-pyrrol-1-yl]acetamide

  • MDL number MFCD06660546
  • Cas Number : 847934-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13866
Availability details for
Quantity
  • Availability
  • Add to Cart