ENA061488178 Enamine

1-(4-butylphenyl)-2-methylpropan-1-amine

  • MDL number MFCD07325242
  • Cas Number : 847837-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13870
Availability details for
Quantity
  • Availability
  • Add to Cart