ENA061488181 Enamine

4-amino-N-(2,2,2-trifluoroethyl)benzene-1-sulfonamide

  • MDL number MFCD07323956
  • Cas Number : 389605-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13874
Availability details for
Quantity
  • Availability
  • Add to Cart