ENA061488182 Enamine

6-methoxyimidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD06660547
  • Cas Number : 180688-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-114
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13875
Availability details for
Quantity
  • Availability
  • Add to Cart