ENA061488186 Enamine

N-(3-chloroquinoxalin-2-yl)thiophene-2-sulfonamide

  • MDL number MFCD07322883
  • Cas Number : 848052-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-177
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13881
Availability details for
Quantity
  • Availability
  • Add to Cart