ENA061488188 Enamine

methyl 4-(2-fluorophenyl)-2,4-dioxobutanoate

  • MDL number MFCD07323611
  • Cas Number : 608536-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13884
Availability details for
Quantity
  • Availability
  • Add to Cart