ENA061488189 Enamine

1-(4-chlorobenzyl)-6-(2-furyl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD06660548
  • Cas Number : 848052-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13887
Availability details for
Quantity
  • Availability
  • Add to Cart