ENA061488192 Enamine

3-amino-N,N,4,5-tetramethylbenzene-1-sulfonamide

  • MDL number MFCD07324910
  • Cas Number : 848179-08-8
  • Molecular Weight : 228.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13894
Availability details for
Quantity
  • Availability
  • Add to Cart