ENA061488193 Enamine

4-{[(aminocarbonyl)amino]methyl}benzoic acid

  • MDL number MFCD07324841
  • Cas Number : 2372-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 291...293
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13900
Availability details for
Quantity
  • Availability
  • Add to Cart