ENA061488197 Enamine

methyl 4-(2,5-dichlorophenyl)-2,4-dioxobutanoate

  • MDL number MFCD07324742
  • Cas Number : 848052-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-153
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13907
Availability details for
Quantity
  • Availability
  • Add to Cart