ENA061488199 Enamine

2-(oxolan-2-yl)acetonitrile

  • MDL number MFCD03206782
  • Cas Number : 33414-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13912
Availability details for
Quantity
  • Availability
  • Add to Cart