ENA061488206 Enamine

5-ethoxy-4-methoxy-2-nitrobenzoic acid

  • MDL number MFCD07323644
  • Cas Number : 61948-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222...224
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13920

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13920
Availability details for
Quantity
  • Availability
  • Add to Cart