ENA061488208 Enamine

1-[(2,3,4-trimethoxyphenyl)methyl]-1H-pyrazol-5-amine

  • MDL number MFCD07323648
  • Cas Number : 956780-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-74
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13923
Availability details for
Quantity
  • Availability
  • Add to Cart