ENA061488222 Enamine

methyl 4-(5-chlorothiophen-2-yl)-2,4-dioxobutanoate

  • MDL number MFCD07324846
  • Cas Number : 848178-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13949
Availability details for
Quantity
  • Availability
  • Add to Cart