ENA061488235 Enamine

3-butyl-8-(chloromethyl)-7-ethyl-3,7-dihydro-1H-purine-2,6-dione

  • MDL number MFCD07324010
  • Cas Number : 848369-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13973
Availability details for
Quantity
  • Availability
  • Add to Cart