ENA061488240 Enamine

N-(3-carbamothioylphenyl)-2-(morpholin-4-yl)acetamide

  • MDL number MFCD07325394
  • Cas Number : 848369-72-2
  • Molecular Weight : 279.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13985
Availability details for
Quantity
  • Availability
  • Add to Cart