ENA061488241 Enamine

N'-(2-cyanoacetyl)thiophene-2-carbohydrazide

  • MDL number MFCD01007924
  • Cas Number : 848369-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13986
Availability details for
Quantity
  • Availability
  • Add to Cart