ENA061488245 Enamine

methyl 4-(2-fluoro-4-methoxyphenyl)-2,4-dioxobutanoate

  • MDL number MFCD07323302
  • Cas Number : 848369-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13990
Availability details for
Quantity
  • Availability
  • Add to Cart