ENA061488247 Enamine

2-(2,5-dimethyl-1H-pyrrol-1-yl)-4-(1-hydroxyethyl)-5-methyl-3-furonitrile

  • MDL number MFCD07325338
  • Cas Number : 848369-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13993

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13993
Availability details for
Quantity
  • Availability
  • Add to Cart