ENA061488251 Enamine

N-(3-carbamothioylphenyl)cyclopropanecarboxamide

  • MDL number MFCD07325397
  • Cas Number : 848369-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14001
Availability details for
Quantity
  • Availability
  • Add to Cart