ENA061488256 Enamine

ethyl 6-(chloromethyl)-4-(2-furyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD03030347
  • Cas Number : 585516-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14008

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14008
Availability details for
Quantity
  • Availability
  • Add to Cart