ENA061488259 Enamine

5-amino-3-[(4-methoxyphenyl)amino]-1H-pyrazole-4-carbonitrile

  • MDL number MFCD04070825
  • Cas Number : 112606-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-120
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13648
Availability details for
Quantity
  • Availability
  • Add to Cart