ENA061488260 Enamine

4-(3-ethylphenyl)-5-(morpholin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07312687
  • Cas Number : 854137-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13649
Availability details for
Quantity
  • Availability
  • Add to Cart