ENA061488264 Enamine

2-(3-chlorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD05664421
  • Cas Number : 845885-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13653

Safety Information

signalword Warning
safety_statement 53-16-25-26-29-33-36/37/39-45
msds https://www.enaminestore.com/catalog/EN300-13653
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart