ENA061488266 Enamine

3-(4-ethyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)propanamide

  • MDL number MFCD07312682
  • Cas Number : 854137-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13655
Availability details for
Quantity
  • Availability
  • Add to Cart