ENA061488268 Enamine

3-(methylsulfanyl)-1,2,4-oxadiazol-5-amine

  • MDL number MFCD03419888
  • Cas Number : 55864-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-103
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13657
Availability details for
Quantity
  • Availability
  • Add to Cart