ENA061488280 Enamine

4-[(4-methylbenzenesulfonyl)oxy]-1-(2-phenyl-2H-1,2,3-triazol-4-yl)butane-1,2,3-triol

  • MDL number MFCD06660544
  • Cas Number : 99099-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13669
Availability details for
Quantity
  • Availability
  • Add to Cart