ENA061488288 Enamine

2-(1-chloroethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole

  • MDL number MFCD07313934
  • Cas Number : 854137-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13677

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13677
Availability details for
Quantity
  • Availability
  • Add to Cart