ENA061488290 Enamine

2-[6-(methoxycarbonyl)-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-5-yl]acetic acid

  • MDL number MFCD07312765
  • Cas Number : 854137-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13679
Availability details for
Quantity
  • Availability
  • Add to Cart