ENA061488300 Enamine

2-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)-2-oxoacetamide

  • MDL number MFCD07310993
  • Cas Number : 956780-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13689

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13689
Availability details for
Quantity
  • Availability
  • Add to Cart