ENA061488301 Enamine

2-(2,5-dimethoxybenzenesulfonamido)acetic acid

  • MDL number MFCD06008335
  • Cas Number : 781656-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13690
Availability details for
Quantity
  • Availability
  • Add to Cart