ENA061488306 Enamine

methyl 4-(2,4-dimethylphenyl)-2,4-dioxobutanoate

  • MDL number MFCD06740813
  • Cas Number : 152034-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13695
Availability details for
Quantity
  • Availability
  • Add to Cart