ENA061488322 Enamine

6-nitro-1,3-benzodioxol-5-amine

  • MDL number MFCD00184722
  • Cas Number : 64993-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13711

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13711
Availability details for
Quantity
  • Availability
  • Add to Cart