ENA061488326 Enamine

4,5-dimethoxy-2-nitroaniline

  • MDL number MFCD00087550
  • Cas Number : 7595-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13715
Availability details for
Quantity
  • Availability
  • Add to Cart