ENA061488346 Enamine

4-(2-chlorophenyl)-5-(pyrrolidin-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07311125
  • Cas Number : 847783-70-4
  • Molecular Weight : 280.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13735
Availability details for
Quantity
  • Availability
  • Add to Cart