ENA061488348 Enamine

2-chloro-5-[methoxy(methyl)sulfamoyl]benzoic acid

  • MDL number MFCD06655895
  • Cas Number : 847758-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13502
Availability details for
Quantity
  • Availability
  • Add to Cart