ENA061488349 Enamine

5-(propylamino)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD07314187
  • Cas Number : 125908-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13738
Availability details for
Quantity
  • Availability
  • Add to Cart