ENA061488350 Enamine

5-(4-fluorophenyl)-4-[2-(morpholin-4-yl)ethyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07312999
  • Cas Number : 847783-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13739
Availability details for
Quantity
  • Availability
  • Add to Cart