ENA061488351 Enamine

4-[(furan-2-yl)methyl]-5-(1H-indol-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07312996
  • Cas Number : 847783-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13740
Availability details for
Quantity
  • Availability
  • Add to Cart