ENA061488354 Enamine

2-[3-(4-ethoxyphenyl)-5-sulfanyl-4H-1,2,4-triazol-4-yl]acetic acid

  • MDL number MFCD07313048
  • Cas Number : 847783-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13743
Availability details for
Quantity
  • Availability
  • Add to Cart