ENA061488355 Enamine

3-[3-(4-ethoxyphenyl)-5-sulfanyl-4H-1,2,4-triazol-4-yl]propanoic acid

  • MDL number MFCD07310904
  • Cas Number : 847783-64-6
  • Molecular Weight : 293.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13744
Availability details for
Quantity
  • Availability
  • Add to Cart