ENA061488382 Enamine

(7-bromo-2,3-dihydro-1,4-benzodioxin-6-yl)acetic acid

  • MDL number MFCD02664762
  • Cas Number : 98947-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13513

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13513
Availability details for
Quantity
  • Availability
  • Add to Cart