ENA061488396 Enamine

ethyl 3-(bromomethyl)-4-fluoro-1-benzothiophene-2-carboxylate

  • MDL number MFCD06655904
  • Cas Number : 854357-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-162
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13520
Availability details for
Quantity
  • Availability
  • Add to Cart