ENA061488408 Enamine

2,4-dimethyl 5-(2-chloroacetamido)-3-(2-methoxy-2-oxoethyl)thiophene-2,4-dicarboxylate

  • MDL number MFCD06655905
  • Cas Number : 854357-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13523
Availability details for
Quantity
  • Availability
  • Add to Cart